2-Nitro-4-(trifluoromethyl)phenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24895G25
AS24895G5
ACTIAS24895G25EA
0
GBP
ACTIAS24895G25
ACTIAS24895G5
2-Nitro-4-(trifluoromethyl)phenol
2-Nitro-4-(trifluoromethyl)phenol
Formula:
C₇H₄F₃NO₃ |
MDL Number:
MFCD00009791 CAS Number: 400-99-7 EINECS: 206-927-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...