2-Fluoro-3-nitro-5-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-5-methyl-3-nitropyridine, 2-Fluoro-3-nitro-5-methylpyridine

AS25234G25 AS25234G5
ACTIAS25234G25EA 236 GBP
ACTIAS25234G25 ACTIAS25234G5
2-Fluoro-3-nitro-5-picoline
2-Fluoro-3-nitro-5-picoline
Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD09475421
CAS Number: 19346-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR