2,4-Difluoro-5-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25245G100 AS25245G25 AS25245G5
ACTIAS25245G100EA 236 GBP
ACTIAS25245G100 ACTIAS25245G25 ACTIAS25245G5
2,4-Difluoro-5-nitrobenzoic acid
2,4-Difluoro-5-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MDL Number: MFCD04115713
CAS Number: 153775-33-8
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR