Methyl 3-fluoro-4-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22693G100 AS22693G25 AS22693G5
ACTIAS22693G100EA 242 GBP
ACTIAS22693G100 ACTIAS22693G25 ACTIAS22693G5
Methyl 3-fluoro-4-nitrobenzoate
Methyl 3-fluoro-4-nitrobenzoate
Formula: C₈H₆FNO₄
MDL Number: MFCD08444027
CAS Number: 185629-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR