3-Fluoro-4-methoxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24816G100 AS24816G25
ACTIAS24816G100EA 232 GBP
ACTIAS24816G100 ACTIAS24816G25
3-Fluoro-4-methoxybenzonitrile
3-Fluoro-4-methoxybenzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD00215835
CAS Number: 331-62-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR