4-(Trifluoromethyl)phenylacetonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Trifluoromethyl)benzyl cyanide

AS24065G100 AS24065G25
ACTIAS24065G100EA 0 GBP
ACTIAS24065G100 ACTIAS24065G25
4-(Trifluoromethyl)phenylacetonitrile
4-(Trifluoromethyl)phenylacetonitrile
Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 131…132 °C (20 mmHg)
Melting Pt: 47.85…48.85 °C
Density: 1.535 g/cm³ (–150.15 °C)
MDL Number: MFCD00001921
CAS Number: 2338-75-2
EINECS: 219-052-2
UN: 3439
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR