5-Fluoro-m-anisaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Fluoro-5-methoxybenzaldehyde, 5-Fluoro-3-methoxybenzaldehyde, 3-Fluoro-5-anisaldehyde
AS33876G1
AS33876G5
ACTIAS33876G1EA
0
GBP
ACTIAS33876G1
ACTIAS33876G5
5-Fluoro-m-anisaldehyde
5-Fluoro-m-anisaldehyde
Formula:
C₈H₇FO₂ MW: 154.14 g/mol |
MDL Number:
MFCD08741376 CAS Number: 699016-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...