(4-Nitrophenyl)acetic acid ethyl ester
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl 4-nitrohenylacetate, Ethyl (4-nitrophenyl)acetate, Ethyl 4-nitrophenylacetate
AS20751G100
AS20751G25
ACTIAS20751G100EA
0
GBP
ACTIAS20751G100
ACTIAS20751G25
(4-Nitrophenyl)acetic acid ethyl ester
(4-Nitrophenyl)acetic acid ethyl ester
Formula:
C₁₀H₁₁NO₄ MW: 209.2 g/mol |
MDL Number:
MFCD00017046 CAS Number: 5445-26-1 EINECS: 226-646-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...