2-Methylbenzamidine hydrochloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methylphenylamidine hydrochloride, 2-Methylbenzene-1-carboximidamide hydrochloride, 2-Methylbenzamidine Hydrochloride

AS21118G1 AS21118G5
ACTIAS21118G1EA 162 GBP
ACTIAS21118G1 ACTIAS21118G5
2-Methylbenzamidine hydrochloride
2-Methylbenzamidine hydrochloride
Formula: C₈H₁₁ClN₂
MW: 170.64 g/mol
MDL Number: MFCD04114417
CAS Number: 18636-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR