Benzamidine hydrochloride hydrate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS25304G100 AS25304G25 AS25304G5
ACTIAS25304G100EA 66 GBP
ACTIAS25304G100 ACTIAS25304G25 ACTIAS25304G5
Benzamidine hydrochloride hydrate
Benzamidine hydrochloride hydrate
Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR