2-Chloro-4-methyl-5-nitropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21282G25 AS21282G5
ACTIAS21282G25EA 0 GBP
ACTIAS21282G25 ACTIAS21282G5
2-Chloro-4-methyl-5-nitropyridine
2-Chloro-4-methyl-5-nitropyridine
Formula: C₆H₅ClN₂O₂
MDL Number: MFCD00010688
CAS Number: 23056-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR