3-Amino-5-chloropyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Chloropyridin-3-amine
AS30566G1
AS30566G5
ACTIAS30566G1EA
0
GBP
ACTIAS30566G1
ACTIAS30566G5
3-Amino-5-chloropyridine
3-Amino-5-chloropyridine
Formula:
C₅H₅ClN₂ |
MDL Number:
MFCD03701386 CAS Number: 22353-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...