4-Chloro-3-(trifluoromethyl)phenol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-5-hydroxybenzotrifluoride
AS24899G100
AS24899G25
ACTIAS24899G100EA
280
GBP
ACTIAS24899G100
ACTIAS24899G25
4-Chloro-3-(trifluoromethyl)phenol
4-Chloro-3-(trifluoromethyl)phenol
Formula:
C₇H₄ClF₃O MW: 196.56 g/mol |
MDL Number:
MFCD00042525 CAS Number: 6294-93-5 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...