α-Chloro-4-(trifluoromethoxy)toluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(Chloromethyl)-4-(trifluoromethoxy)benzene, 4-(Trifluoromethoxy)benzyl chloride, p-(Trifluoromethoxy)benzyl chloride, 1-(Chlormethyl)-4-(trifluormethoxy)benzene

AS24339G100 AS24339G25 AS24339G5
ACTIAS24339G100EA 0 GBP
ACTIAS24339G100 ACTIAS24339G25 ACTIAS24339G5
α-Chloro-4-(trifluoromethoxy)toluene
α-Chloro-4-(trifluoromethoxy)toluene
Formula: C₈H₆ClF₃O
MDL Number: MFCD00052326
CAS Number: 65796-00-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR