4'-Chloro-2',6'-difluoroacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(4-Chloro-2,6-difluorophenyl)ethanone
AS35474G1
AS35474G5
ACTIAS35474G1EA
0
GBP
ACTIAS35474G1
ACTIAS35474G5
4'-Chloro-2',6'-difluoroacetophenone
4'-Chloro-2',6'-difluoroacetophenone
Formula:
C₈H₅ClF₂O MW: 190.58 g/mol |
MDL Number:
MFCD09832259 CAS Number: 1017777-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...