3-Chlorothiobenzamide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20696G25 AS20696G5
ACTIAS20696G25EA 0 GBP
ACTIAS20696G25 ACTIAS20696G5
3-Chlorothiobenzamide
3-Chlorothiobenzamide
Formula: C₇H₆ClNS
MDL Number: MFCD00276993
CAS Number: 2548-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR