3-Chloro-5-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21388G1 AS21388G5
ACTIAS21388G1EA 0 GBP
ACTIAS21388G1 ACTIAS21388G5
3-Chloro-5-nitrobenzoic acid
3-Chloro-5-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD09263224
CAS Number: 34662-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR