2-Amino-4-bromopyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS50968G1
AS50968G5
ACTIAS50968G1EA
0
GBP
ACTIAS50968G1
ACTIAS50968G5
2-Amino-4-bromopyridine
2-Amino-4-bromopyridine
Formula:
C₅H₅BrN₂ |
MDL Number:
MFCD01646115 CAS Number: 84249-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...