3-Bromo-5-fluorophenol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-3-fluoro-5-hydroxybenzene

AS22765G100 AS22765G25
ACTIAS22765G100EA 0 GBP
ACTIAS22765G100 ACTIAS22765G25
3-Bromo-5-fluorophenol
3-Bromo-5-fluorophenol
Formula: C₆H₄BrFO
MW: 191 g/mol
MDL Number: MFCD07783710
CAS Number: 433939-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR