3-Bromo-2-fluoro-6-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-2-fluoro-6-picoline

AS31830G1 AS31830G5
ACTIAS31830G1EA 0 GBP
ACTIAS31830G1 ACTIAS31830G5
3-Bromo-2-fluoro-6-methylpyridine
3-Bromo-2-fluoro-6-methylpyridine
Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03095080
CAS Number: 375368-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR