Methyl 4-bromo-2-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23488G25 AS23488G5
ACTIAS23488G25EA 0 GBP
ACTIAS23488G25 ACTIAS23488G5
Methyl 4-bromo-2-nitrobenzoate
Methyl 4-bromo-2-nitrobenzoate
Formula: C₈H₆BrNO₄
MDL Number: MFCD10699645
CAS Number: 158580-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR