Methyl 3-bromo-5-nitrobenzoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22711G1
AS22711G25
AS22711G5
ACTIAS22711G1EA
0
GBP
ACTIAS22711G1
ACTIAS22711G25
ACTIAS22711G5
Methyl 3-bromo-5-nitrobenzoate
Methyl 3-bromo-5-nitrobenzoate
Formula:
C₈H₆BrNO₄ |
MDL Number:
MFCD09258759 CAS Number: 6307-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...