Ethyl 3-bromo-5-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 3-bromo-5-nitrobenzoate, 3-Bromobenzoic acid ethyl ester

AS22710G25 AS22710G5
ACTIAS22710G25EA 0 GBP
ACTIAS22710G25 ACTIAS22710G5
Ethyl 3-bromo-5-nitrobenzoate
Ethyl 3-bromo-5-nitrobenzoate
Formula: C₉H₈BrNO₄
MDL Number: MFCD09258760
CAS Number: 690260-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR