(4-Bromophenyl)acetic acid ethyl ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-(4-bromophenyl)acetate, Ethyl (4-bromophenyl)acetate

AS20470G100 AS20470G25
ACTIAS20470G100EA 0 GBP
ACTIAS20470G100 ACTIAS20470G25
(4-Bromophenyl)acetic acid ethyl ester
(4-Bromophenyl)acetic acid ethyl ester
Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD00016333
CAS Number: 14062-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR