4-Bromo-2-chlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23083G100 AS23083G25 AS23083G5
ACTIAS23083G100EA 193 GBP
ACTIAS23083G100 ACTIAS23083G25 ACTIAS23083G5
4-Bromo-2-chlorobenzonitrile
4-Bromo-2-chlorobenzonitrile
Formula: C₇H₃BrClN
MW: 216.46 g/mol
Boiling Pt: 142…143 °C (10 mmHg)
Melting Pt: 67…68 °C
MDL Number: MFCD00040883
CAS Number: 154607-01-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR