3-Bromo-4-chlorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24906G100 AS24906G25 AS24906G5
ACTIAS24906G100EA 0 GBP
ACTIAS24906G100 ACTIAS24906G25 ACTIAS24906G5
3-Bromo-4-chlorobenzonitrile
3-Bromo-4-chlorobenzonitrile
Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD09025661
CAS Number: 948549-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR