1,3-Dibromo-2,2-dimethoxypropane

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21649G100 AS21649G25
ACTIAS21649G100EA 0 GBP
ACTIAS21649G100 ACTIAS21649G25
1,3-Dibromo-2,2-dimethoxypropane
1,3-Dibromo-2,2-dimethoxypropane
Formula: BrH₂CC(OCH₃)₂CH₂Br
MDL Number: MFCD00051792
CAS Number: 22094-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR