2,5-Dichlorobenzophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2,5-Dichlorophenyl)(phenyl)methanone

AS24920G100 AS24920G25
ACTIAS24920G100EA 0 GBP
ACTIAS24920G100 ACTIAS24920G25
2,5-Dichlorobenzophenone
2,5-Dichlorobenzophenone
Formula: C₁₃H₈Cl₂O
MDL Number: MFCD00079746
CAS Number: 16611-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR