7-Chloroquinaldine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24452G100 AS24452G25
ACTIAS24452G100EA 0 GBP
ACTIAS24452G100 ACTIAS24452G25
7-Chloroquinaldine
7-Chloroquinaldine
Formula: C₁₀H₈ClN
MDL Number: MFCD00075436
CAS Number: 4965-33-7
EINECS: 440-600-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR