4-Fluoro-2-(trifluoromethyl)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Cyano-5-fluorobenzotrifluoride

AS20031G100 AS20031G25
ACTIAS20031G100EA 0 GBP
ACTIAS20031G100 ACTIAS20031G25
4-Fluoro-2-(trifluoromethyl)benzonitrile
4-Fluoro-2-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 52…55 °C (15 mmHg)
Melting Pt: 43…45 °C
Density: 1.35 g/cm³ (20 °C)
MDL Number: MFCD00061283
CAS Number: 194853-86-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR