2-Fluoro-4-(trifluoromethyl)benzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24244G25
AS24244G5
ACTIAS24244G25EA
0
GBP
ACTIAS24244G25
ACTIAS24244G5
2-Fluoro-4-(trifluoromethyl)benzonitrile
2-Fluoro-4-(trifluoromethyl)benzonitrile
Formula:
C₈H₃F₄N Boiling Pt: 178 °C (1013 hPa) Density: 1.35 g/cm³ (20 °C) |
MDL Number:
MFCD00061279 CAS Number: 146070-34-0 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...