L-Phenylalaninol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21489G100 AS21489G25
ACTIAS21489G100EA 0 GBP
ACTIAS21489G100 ACTIAS21489G25
L-Phenylalaninol
L-Phenylalaninol
Formula: C₉H₁₃NO
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 91…94 °C
MDL Number: MFCD00004732
CAS Number: 3182-95-4
EINECS: 221-674-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR