4-(Methylsulphonyl)benzyl bromide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: α-Bromo-4-(methylsulphonyl)toluene, 1-(Bromomethyl)-4-(methylsulphonyl)benzene

AS20402G1 AS20402G25 AS20402G5
ACTIAS20402G1EA 29 GBP
ACTIAS20402G1 ACTIAS20402G25 ACTIAS20402G5
4-(Methylsulphonyl)benzyl bromide
4-(Methylsulphonyl)benzyl bromide
Formula: C₈H₉BrO₂S
MW: 249.13 g/mol
Melting Pt: 97 °C
MDL Number: MFCD00185849
CAS Number: 53606-06-7
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR