Methyl-5-chloro-2-nitrobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23116G100 AS23116G25
ACTIAS23116G100EA 0 GBP
ACTIAS23116G100 ACTIAS23116G25
Methyl-5-chloro-2-nitrobenzoate
Methyl 5-chloro-2-nitrobenzoate
Formula: C₈H₆ClNO₄
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 48.5 °C
Density: 1,426 g/cm³ (20 °C)
MDL Number: MFCD00272139
CAS Number: 51282-49-6
EINECS: 257-107-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR