3-Pyridineboronic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Pyridinylboronic acid, Pyridin-3-ylboronic acid, Pyridine-3-boronic acid
AS50987G25
AS50987G5
ACTIAS50987G25EA
0
GBP
ACTIAS50987G25
ACTIAS50987G5
3-Pyridineboronic acid
3-Pyridineboronic acid
Formula:
C₅H₆BNO₂ |
MDL Number:
MFCD00674177 CAS Number: 1692-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...