5-Amino-2-fluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23139G100 AS23139G25 AS23139G5
ACTIAS23139G100EA 0 GBP
ACTIAS23139G100 ACTIAS23139G25 ACTIAS23139G5
5-Amino-2-fluorobenzonitrile
5-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
MDL Number: MFCD00277872
CAS Number: 53312-81-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR