4-Amino-3-bromobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS25375G100
AS25375G25
ACTIAS25375G100EA
0
GBP
ACTIAS25375G100
ACTIAS25375G25
4-Amino-3-bromobenzonitrile
Formula:
C₇H₅BrN₂ |
MDL Number:
MFCD01861386 CAS Number: 50397-74-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...