2-Iodobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
o-Iodobenzonitrile
AS21594G100
AS21594G25
ACTIAS21594G100EA
0
GBP
ACTIAS21594G100
ACTIAS21594G25
2-Iodobenzonitrile
2-Iodobenzonitrile
Formula:
C₇H₄IN MW: 229.02 g/mol |
MDL Number:
MFCD00079761 CAS Number: 4387-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...