2-Amino-3-chloro-5-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-chloro-6-cyano-4-nitroaniline

AS21381G100 AS21381G25
ACTIAS21381G100EA 127 GBP
ACTIAS21381G100 ACTIAS21381G25
2-Amino-3-chloro-5-nitrobenzonitrile
2-Amino-3-chloro-5-nitrobenzonitrile
Formula: C₇H₄ClN₃O₂
MW: 197.58 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD00126957
CAS Number: 20352-84-5
EINECS: 243-760-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR