(2-Nitrophenyl)acetonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS22261G100
AS22261G25
ACTIAS22261G100EA
0
GBP
ACTIAS22261G100
ACTIAS22261G25
(2-Nitrophenyl)acetonitrile
(2-Nitrophenyl)acetonitrile
Formula:
C₈H₆N₂O₂ Boiling Pt: 304 °C (1013 hPa) Melting Pt: 81…83 °C |
MDL Number:
MFCD00007183 CAS Number: 610-66-2 EINECS: 210-231-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...