6-Methylnicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Methyl-3-pyridinecarboxylic acid, 6-Methylpyridine-3-carboxylic acid

AS20173G100 AS20173G25
ACTIAS20173G100EA 0 GBP
ACTIAS20173G100 ACTIAS20173G25
6-Methylnicotinic acid
6-Methylnicotinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 212
MDL Number: MFCD00006341
CAS Number: 3222-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR