5-Methylnicotinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Methyl-3-pyridinecarboxylic acid, 5-Methylpyridine-3-carboxylic acid
AS21378G25
AS21378G5
ACTIAS21378G25EA
32
GBP
ACTIAS21378G25
ACTIAS21378G5
5-Methylnicotinic acid
5-Methylnicotinic acid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol |
MDL Number:
MFCD00829036 CAS Number: 3222-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...