Piperidin-4-ol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Piperidinol, 4-Hydroxypiperidine

AS20159G100 AS20159G25
ACTIAS20159G100EA 0 GBP
ACTIAS20159G100 ACTIAS20159G25
Piperidin-4-ol
Piperidin-4-ol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 222
Melting Pt: 86…89 °C
Flash Pt: <100 °C
MDL Number: MFCD00005999
CAS Number: 5382-16-1
EINECS: 226-373-1
UN: 3263
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR