3-Phenylpiperidine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS36368G1
AS36368G5
ACTIAS36368G1EA
0
GBP
ACTIAS36368G1
ACTIAS36368G5
3-Phenylpiperidine
3-Phenylpiperidine
Formula:
C₁₁H₁₅N MW: 161.25 g/mol |
MDL Number:
MFCD00804860 CAS Number: 3973-62-4 EINECS: 223-602-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...