3-Aminophenylboronic acid monohydrate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate

AS20217G25 AS20217G5
ACTIAS20217G25EA 0 GBP
ACTIAS20217G25 ACTIAS20217G5
3-Aminophenylboronic acid monohydrate
3-Aminophenylboronic acid monohydrate
Formula: C₆H₈BNO₂·1H₂O
MDL Number: MFCD00149554
CAS Number: 206658-89-1
EINECS: 250-189-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR