2-Methylnicotinic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid
AS22538G25
AS22538G5
ACTIAS22538G25EA
98
GBP
ACTIAS22538G25
ACTIAS22538G5
2-Methylnicotinic acid
2-Methylnicotinic acid
Formula:
C₇H₇NO₂ MW: 137.14 g/mol |
MDL Number:
MFCD00013449 CAS Number: 3222-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...