Thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21356G100 AS21356G25
ACTIAS21356G100EA 0 GBP
ACTIAS21356G100 ACTIAS21356G25
Thiazole
Thiazole
Formula: C₃H₃NS
Boiling Pt: 116.8 °C (1013 hPa)
Melting Pt: –34 °C
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00005315
CAS Number: 288-47-1
EINECS: 206-021-3
Merck Index: 12,09442

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR