Tetrakis(triphenylphosphine)palladium(0)

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

AS50331G1 AS50331G25 AS50331G5
ACTIAS50331G1EA 0 GBP
ACTIAS50331G1 ACTIAS50331G25 ACTIAS50331G5
Tetrakis(triphenylphosphine)palladium(0)
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155.58 g/mol
Melting Pt: 103…107 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR