Ethyl propiolate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20126G25 AS20126G5
ACTIAS20126G25EA 0 GBP
ACTIAS20126G25 ACTIAS20126G5
Ethyl propiolate
Ethyl propiolate
Formula: C₅H₆O₂
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: 9 °C
Density: 0.9788 g/cm³ (4 °C)
MDL Number: MFCD00009184
CAS Number: 623-47-2
EINECS: 210-795-8
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR