Indazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20033G100
AS20033G25
AS20033G5
ACTIAS20033G100EA
0
GBP
ACTIAS20033G100
ACTIAS20033G25
ACTIAS20033G5
Indazole
Indazole
Formula:
C₇H₆N₂ Boiling Pt: 270 °C (1013 hPa) Melting Pt: 147…149 °C Density: 1.32 g/cm³ (20 °C) |
MDL Number:
MFCD00005691 CAS Number: 271-44-3 EINECS: 205-978-4 Merck Index: 13,04961 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...